Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

306010

Sigma-Aldrich

3,5-Difluoronitrobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3NO2
CAS Number:
Molecular Weight:
159.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

176-177 °C (lit.)

mp

17 °C (lit.)

density

1.407 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(F)cc(F)c1

InChI

1S/C6H3F2NO2/c7-4-1-5(8)3-6(2-4)9(10)11/h1-3H

InChI key

AUQBBDWDLJSKMI-UHFFFAOYSA-N

General description

Molecular structure of 3,5-difluoronitrobenzene was studied by gas-phase electron diffraction, MP2 ab initio and by B3LYP density functional calculations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga V Dorofeeva et al.
The journal of physical chemistry. A, 112(22), 5002-5009 (2008-05-09)
3,5-Difluoronitrobenzene (3,5-DFNB) and 2,6-difluoronitrobenzene (2,6-DFNB) have been studied by gas-phase electron diffraction (GED), MP2 ab initio, and by B3LYP density functional calculations. Refinements of r h1 and r e static and r h1 dynamic GED models were carried out for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service