Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

259187

Sigma-Aldrich

Hexetidine, mixture of stereoisomers

Synonym(s):

5-Amino-1,3-bis(2-ethylhexyl)hexahydro-5-methylpyrimidine, NSC 17764

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H45N3
CAS Number:
Molecular Weight:
339.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.4649 (lit.)

bp

160 °C/0.4 mmHg (lit.)

solubility

acetone: soluble(lit.)
benzene: soluble(lit.)
chloroform: soluble(lit.)
ethanol: soluble(lit.)
hexane: soluble(lit.)
methanol: soluble(lit.)
petroleum ether: soluble(lit.)
water: insoluble(lit.)

density

0.889 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)CN1CN(CC(CC)CCCC)CC(C)(N)C1

InChI

1S/C21H45N3/c1-6-10-12-19(8-3)14-23-16-21(5,22)17-24(18-23)15-20(9-4)13-11-7-2/h19-20H,6-18,22H2,1-5H3

InChI key

DTOUUUZOYKYHEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The effectiveness of a hexetidine mouthwash in reducing supragingival plaque and gingival inflammation has been examined.

Application

Involved in studies:
  • To identify pure liquid salt forms of aspirin
  • Of alkyl based selective displacers for protein purification via ion exchange chromatography
  • Of the in vivo application of models for testing drugs against nonreplicating Mycobacterium tuberculosis
  • Nutrient-sensitized screening for drugs that shift the energy metabolism from mitochondrial respiration to glycolysis
  • Inhibition of angiogensis
  • Screening antimalarial drugs

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G A Van der Weijden et al.
International journal of dental hygiene, 8(4), 276-279 (2010-10-22)
Unilateral or bilateral swelling of the parotid gland is a reported side effect of rinsing the mouth with chlorhexidine. Although the incidence rate is extremely low, there have been several case reports on this topic and the authors of these
G Giuliana et al.
Oral surgery, oral medicine, oral pathology, oral radiology, and endodontics, 87(1), 44-49 (1999-02-02)
Antimicrobial mouthrinses may represent a valid alternative to topical antifungal agents. However, the action of antimicrobials could be affected by the different ingredients incorporated into mouthrinse products. The purpose of the present study was to investigate the in vitro antifungal
C P McCoy et al.
The Journal of pharmacy and pharmacology, 52(11), 1355-1359 (2001-02-24)
The non-antibiotic antimicrobial agent hexetidine is widely used at a concentration of 0.1% w/v as an oral rinse to reduce the number of viable microorganisms within the oral cavity. However, following use, the available concentration of hexetidine in the oral
F Ruza et al.
European journal of epidemiology, 14(7), 719-727 (1998-12-16)
To assess the effectiveness of selective digestive decontamination (SDD) on the control of nosocomial infection (NI) in critically ill pediatric patients. A prospective, randomized, non-blinded and controlled clinical microbiology study. The pediatric intensive care unit (PICU) of a tertiary level
N C Sharma et al.
Journal of clinical periodontology, 30(7), 590-594 (2003-07-02)
To assess the antiplaque/antigingivitis efficacy of a hexetidine-containing mouthwash. This examiner-blind, parallel group, controlled clinical study examined the effectiveness of a hexetidine (0.1%) mouthwash both in inhibiting the development of supragingival plaque and in reducing gingivitis. One hundred and thirty-four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service