Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

256552

Sigma-Aldrich

Barium titanate(IV)

powder, 99.995% trace metals basis

Synonym(s):

Titanium barium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaTiO3
CAS Number:
Molecular Weight:
233.19
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.995% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: titanium

density

6.08 g/mL at 25 °C (lit.)

SMILES string

[Ba++].[O-][Ti]([O-])=O

InChI

1S/Ba.3O.Ti/q+2;;2*-1;

InChI key

WNKMTAQXMLAYHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyuichi Yasui et al.
Ultrasonics sonochemistry, 18(5), 1211-1217 (2011-04-05)
Numerical simulations of sonochemical production of nanoparticles have been performed for the first time under the experimental condition of Dang et al. [Jpn. J. Appl. Phys. 48 (2009) 09KC02] on the production of BaTiO(3). The results of the numerical simulations
Sung Park et al.
Journal of nanoscience and nanotechnology, 13(5), 3437-3440 (2013-07-19)
A unique synthesis method was developed, which is called solution combustion method (SCM). TiO2 nanopowder was synthesized by this method. This SCM TiO2 nanopowder (-35 nm) was added to the dielectric layer of AC powder electroluminescence (EL) device. The dielectric
R Anand Theerthan et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 24(40), 405901-405901 (2012-09-07)
Artificial tuning of dielectric parameters can result from interface conductivity in polycrystalline materials. In ferroelectric single crystals, it has already been shown that ferroelectric domain walls can be the source of such artificial coupling. We show here that low-temperature dielectric
Hee Han et al.
Advanced materials (Deerfield Beach, Fla.), 23(40), 4599-4613 (2011-09-16)
With the continued demand for ultrahigh density ferroelectric data storage applications, it is becoming increasingly important to scale the dimension of ferroelectrics down to the nanometer-scale region and to thoroughly understand the effects of miniaturization on the materials properties. Upon
Deepam Maurya et al.
Dalton transactions (Cambridge, England : 2003), 41(18), 5643-5652 (2012-03-21)
Nanostructured lead-free ferroelectric Na(0.5)Bi(0.5)TiO(3)-BaTiO(3) (NBTBT) whiskers with a high aspect ratio were synthesized topochemically using Na(2)Ti(6)O(13) (NTO) as a host structure for the first time. High energy X-ray diffraction coupled with an atomic pair distribution function (PDF) and Raman scattering

Articles

Perovskite-phase metal oxides exhibit a variety of interesting physical properties which include ferroelectric, dielectric, pyroelectric, and piezoelectric behavior.

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service