Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

169951

Sigma-Aldrich

Methanesulfonyl chloride

≥99%

Synonym(s):

Mesyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2Cl
CAS Number:
Molecular Weight:
114.55
Beilstein:
506297
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

≥99%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

60 °C/21 mmHg (lit.)

density

1.48 g/mL at 25 °C (lit.)

SMILES string

CS(Cl)(=O)=O

InChI

1S/CH3ClO2S/c1-5(2,3)4/h1H3

InChI key

QARBMVPHQWIHKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazumi Suzuki et al.
Journal of controlled release : official journal of the Controlled Release Society, 301, 28-41 (2019-03-08)
Nanomedicine modification with ligands directed to receptors on tumor blood vessels has the potential for selectively enhancing nanomedicine accumulation in malignant tissues by overcoming the vascular barrier of tumors. Nevertheless, the development of broadly applicable ligand approaches capable of promoting
Giovanna Cutrone et al.
Carbohydrate polymers, 223, 115085-115085 (2019-08-21)
Nanoparticles made of metal-organic frameworks (nanoMOFs) are becoming of increasing interest as drug carriers. However, engineered coatings such as poly(ethylene glycol) (PEG) based ones are required to prevent nanoMOFs recognition and clearance by the innate immune system, a prerequisite for
Sonu Kumar et al.
Macromolecular bioscience, 20(6), e2000048-e2000048 (2020-04-15)
The synthesis and self-assembly of peptide-polymer conjugates into fibrillar nanostructures are reported, based on the amyloidogenic peptide KLVFF. A strategy for rational synthesis of polymer-peptide conjugates is documented via tethering of the amyloidogenic peptide segment LVFF (Aβ17-20 ) and its
Zdzisław Brzozowski et al.
Journal of medicinal chemistry, 45(2), 430-437 (2002-01-11)
A new series of 8-chloro-5,5-dioxoimidazo[1,2-b][1,4,2]benzodithiazine derivatives 8-25 with heteroaryloxycarbonyl or heteroarylcarbamoyl substituents at position 7 have been synthesized as potential antitumor agents. In this procedure a novel type of mixed anhydride 7 was prepared from 8-chloro-5,5-dioxoimidazo[1,2-b][1,4,2]benzodithiazine-7-carboxylic acid 6 and methanesulfonyl
Hiroshi Nagase et al.
The Journal of organic chemistry, 73(20), 8093-8096 (2008-09-25)
The treatment of morphinan 1 with NaH and MsCl provided very stable iminium salt 7 possessing propellane skeleton. One of the synthesized iminium salts 7, isobutyl derivative 7b, was crystallized and its structure was determined by X-ray crystallography. The natural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service