Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

146706

Sigma-Aldrich

4,4′-Dimethylbenzil

97%

Synonym(s):

1,2-Bis(4-methylphenyl)-1,2-ethanedione, 1,2-Bis(4-methylphenyl)ethane-1,2-dione, p ,p ′-Dimethylbenzil, Bis(4-methylphenyl)ethanedione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COCOC6H4CH3
CAS Number:
Molecular Weight:
238.28
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

102-104 °C (lit.)

SMILES string

Cc1ccc(cc1)C(=O)C(=O)c2ccc(C)cc2

InChI

1S/C16H14O2/c1-11-3-7-13(8-4-11)15(17)16(18)14-9-5-12(2)6-10-14/h3-10H,1-2H3

InChI key

BCWCEHMHCDCJAD-UHFFFAOYSA-N

Gene Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R de la Fuente et al.
Journal of bioluminescence and chemiluminescence, 7(1), 27-35 (1992-01-01)
The pyrolyisis of di-tert-butyl peroxyoxalate in the presence of para-substituted benzaldehydes produces almost quantitatively the corresponding p,p'-disubstituted benzils. The formation of these products is accompanied by chemiluminescence arising from excited triplets. From the quantum yield of excited triplet generation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service