Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

144606

Sigma-Aldrich

Cyanomethyl benzenesulfonate

80%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO3CH2CN
CAS Number:
Molecular Weight:
197.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

80%

refractive index

n20/D 1.523 (lit.)

bp

142-146 °C/1 mmHg (lit.)

density

1.309 g/mL at 25 °C (lit.)

functional group

nitrile
sulfonic acid

SMILES string

O=S(=O)(OCC#N)c1ccccc1

InChI

1S/C8H7NO3S/c9-6-7-12-13(10,11)8-4-2-1-3-5-8/h1-5H,7H2

InChI key

SSOVHFPWMPUQOS-UHFFFAOYSA-N

General description

Cyanomethyl benzenesulfonate is a potential cyanomethylating agent.

Application

Cyanomethyl benzenesulfonate was used in the synthesis of nido-and closo-arsacarboranes. It was used in separation of aromatic hydrophobic sulfonates by micellar electrokinetic chromatography using different buffer system.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of nido-and closo-arsacarboranes.
Smith J, et al.
Inorganic Chemistry, 13(10), 2312-2316 (1974)
[Studies on application of cyanomethyl benzenesulfonate as a new cyanomethylating agent. I. Cyanomethyl esters of arylsulfonic acids].
S Grudziński
Acta poloniae pharmaceutica, 23(5), 417-423 (1966-01-01)
Sille Ehala et al.
Journal of chromatography. A, 1161(1-2), 322-326 (2007-07-14)
Two different buffer systems for the separation of 12 aromatic hydrophobic sulfonates by micellar electrokinetic chromatography (MEKC) were developed. The following buffer systems were used: aqueous phosphate buffers containing either cetyltrimethylammonium bromide (CTAB) or sodium dodecyl sulfate (SDS). Eleven aromatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service