Merck
  • Bevacizumab increases viral distribution in human anaplastic thyroid carcinoma xenografts and enhances the effects of E1A-defective adenovirus dl922-947.
Bevacizumab increases viral distribution in human anaplastic thyroid carcinoma xenografts and enhances the effects of E1A-defective adenovirus dl922-947.
Clinical cancer research : an official journal of the American Association for Cancer Research (2008-10-18)
Silvana Libertini, Irma Iacuzzo, Giuseppe Perruolo, Stefania Scala, Caterina Ieranò, Renato Franco, Gunnel Hallden, Giuseppe Portella
RESUMO
Anaplastic thyroid carcinoma is a prime target for innovative therapy because it represents one of the most lethal human neoplasms and is refractory to conventional treatments such as chemotherapy and radiotherapy. We have evaluated a novel therapeutic approach based on the oncolytic replication-selective adenovirus dl922-947. The antitumor efficacies of the E1ADeltaCR2 (dl922-947) and DeltaE1B55K (dl1520) mutants were compared in human thyroid anaplastic carcinoma cells in culture and in xenografts in vivo. To enhance the effects of dl922-947, anaplastic thyroid carcinoma tumor xenografts were treated with dl922-947 in combination with bevacizumab. We showed that the efficacy of dl922-947 exceeded that of dl1520 in all tested anaplastic thyroid carcinoma cells in vitro and in vivo. Furthermore, bevacizumab in combination with dl922-947 significantly reduced tumor growth compared with single treatments alone. Bevacizumab treatment significantly improved viral distribution in neoplastic tissues. Our data showed that dl922-947 had a higher oncolytic activity compared with dl1520 in anaplastic thyroid carcinoma cell lines and might represent a better option for virotherapy of anaplastic thyroid carcinoma. Moreover, bevacizumab increased the oncolytic effects of dl922-947 by enhancing viral distribution in tumors. The results described herein encourage the use of the dl922-947 virus in combination with bevacizumab.