Merck
  • Nitric oxide biosynthesis by Leishmania amazonensis promastigotes containing a high percentage of metacyclic forms.
Nitric oxide biosynthesis by Leishmania amazonensis promastigotes containing a high percentage of metacyclic forms.
Archives of microbiology (2006-04-01)
Marcelo Genestra, Wilson J S Souza, Damiana Guedes-Silva, Gérzia M C Machado, Léa Cysne-Finkelstein, Rômulo José Soares Bezerra, Fabiane Monteiro, Leonor L Leon
RESUMO
Due to the diversity of its physiological and pathophysiological functions and general ubiquity, the study of nitric oxide (NO) has become of great interest. In this work, it was demonstrated that Leishmania amazonensis promastigotes produces NO, a free radical synthesized from L: -arginine by nitric oxide synthase (NOS). A soluble NOS was purified from L. amazonensis promastigotes by affinity chromatography (2', 5'-ADP-agarose) and on SDS-PAGE the enzyme migrates as a single protein band of 116.2 (+/-6) kDa. Furthermore, the presence of a constitutive NOS was detected through indirect immunofluorescence using anti-cNOS and in NADPH consumption assays. The present work show that NO production, detected as nitrite in culture supernatant, is prominent in promastigotes preparations with high number of metacyclic forms, suggesting an association with the differentiation and the infectivity of the parasite.