Merck
Hydrogen sulfide is a novel prosecretory neuromodulator in the Guinea-pig and human colon.
Gastroenterology (2006-11-15)
Rudolf Schicho, Dagmar Krueger, Florian Zeller, Claus Werner Hann Von Weyhern, Thomas Frieling, Hideo Kimura, Isao Ishii, Roberto De Giorgio, Barbara Campi, Michael Schemann
RESUMO
Hydrogen sulfide (H(2)S) has been suggested as a novel gasomediator. We explored its unknown neuromodulatory role in human and guinea-pig colon. We used immunohistochemistry to detect H(2)S-producing enzymes cystathionine gamma-lyase (CSE) and cystathionine beta-synthase (CBS) in enteric neurons, Ussing chambers to measure mucosal ion secretion, and neuroimaging with voltage- and Ca(++)-sensitive dyes to record H(2)S effects on guinea-pig and human enteric neurons. More than 90% of guinea-pig and human submucous and myenteric neurons were colabeled for CSE and CBS. Myenteric interstitial cells of Cajal were CSE-immunoreactive. The exogenous H(2)S donor NaHS (0.2-2.5 mmol/L) concentration-dependently increased chloride secretion in human and guinea-pig submucosa/mucosa preparations, but not in the colonic epithelial cell line T84. The secretory response was reduced significantly by tetrodotoxin (0.5 micromol/L), capsaicin desensitization (10 micromol/L), and the transient receptor potentials vanilloid receptor 1 antagonist capsazepine (10 micromol/L). The endogenous H(2)S donor L-cysteine also induced secretion that was diminished significantly by capsaicin desensitization, the CBS inhibitor amino-oxyacetic acid, and the CSE inhibitor propargylglycine. NaHS increased spike discharge in 23% of guinea-pig and 36% of human submucous neurons, but had no effect on Ca(++) mobilization in cultured guinea-pig enteric neurons. This excitatory response was reduced significantly by capsaicin desensitization and capsazepine, but not by glibenclamide (10 micromol/L). The presence of H(2)S-producing enzymes in human and guinea-pig enteric neurons, the excitatory action on enteric neurons, and the prosecretory effects of NaHS suggest H(2)S as a novel gut-signaling molecule. Its action mainly involves transient receptor potentials vanilloid receptor 1 receptors on extrinsic afferent terminals, which in turn activate enteric neurons.