Merck
Evidence for estrogen receptor alpha and beta expression in skeletal muscle of pigs.
Histochemistry and cell biology (2006-08-10)
Claudia Kalbe, Marcus Mau, Karin Wollenhaupt, Charlotte Rehfeldt
RESUMO
Recent research suggests that estrogen receptors (ERs) are of significance in skeletal muscle function. The aim of the present study was to investigate, whether ERalpha and ERbeta are expressed in different porcine skeletal muscles and in satellite cells derived from semimembranosus muscle (SM) at the protein and mRNA level. Immunohistochemistry demonstrated positive staining for ERalpha in the nuclei of skeletal muscle cells, while the ERbeta stain showed positive signals in nuclei and cytoplasm of skeletal myofibers and myoblasts derived from satellite cells. Additionally, a weak expression of both ER subtypes was seen in skeletal muscle tissue and SM satellite cells with Western blot analysis. A clear expression of the ERalpha mRNA and a weak expression of the ERbeta mRNA was seen in skeletal muscle tissue and SM satellite cell cultures, as determined by reverse transcription (RT)-PCR. The present study shows for the first time that both ERalpha and ERbeta are expressed in porcine skeletal muscle, which, consequently, could be considered as a target tissue for estrogens or estrogen-like compounds. However, more detailed studies on the functional impact of both receptor subtypes in skeletal muscle are necessary. The porcine SM satellite cell culture provides a suitable in vitro model to investigate estrogenic effects on pig skeletal muscle.