Merck
  • Oxidative status of DJ-1-dependent activation of dopamine synthesis through interaction of tyrosine hydroxylase and 4-dihydroxy-L-phenylalanine (L-DOPA) decarboxylase with DJ-1.
Oxidative status of DJ-1-dependent activation of dopamine synthesis through interaction of tyrosine hydroxylase and 4-dihydroxy-L-phenylalanine (L-DOPA) decarboxylase with DJ-1.
The Journal of biological chemistry (2009-08-26)
Shizuma Ishikawa, Takahiro Taira, Takeshi Niki, Kazuko Takahashi-Niki, Chinatsu Maita, Hiroshi Maita, Hiroyoshi Ariga, Sanae M M Iguchi-Ariga
RESUMO
Parkinson disease (PD) is caused by loss of dopamine, which is synthesized from tyrosine by two enzymes, tyrosine hydroxylase (TH) and 4-dihydroxy-L-phenylalanine decarboxylase (DDC). DJ-1 is a causative gene for the familial form of PD, but little is known about the roles of DJ-1 in dopamine synthesis. In this study, we found that DJ-1 directly bound to TH and DDC and positively regulated their activities in human dopaminergic cells. Mutants of DJ-1 found in PD patients, including heterozygous mutants, lost their activity and worked as dominant-negative forms toward wild-type DJ-1. When cells were treated with H(2)O(2), 6-hydroxydopamine, or 1-methyl-4-phenylpyridinium, changes in activities of TH and DDC accompanied by oxidation of cysteine 106 of DJ-1 occurred. It was found that DJ-1 possessing Cys-106 with SH and SOH forms was active and that DJ-1 possessing Cys-106 with SO(2)H and SO(3)H forms was inactive in terms of stimulation of TH and DDC activities. These findings indicate an essential role of DJ-1 in dopamine synthesis and contribution of DJ-1 to the sporadic form of PD.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
ANTI-FLAG® M2 monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)