Merck
  • Clinical features of human papilloma virus-related head and neck squamous cell carcinoma of an unknown primary site.
Clinical features of human papilloma virus-related head and neck squamous cell carcinoma of an unknown primary site.
ORL; journal for oto-rhino-laryngology and its related specialties (2014-07-06)
Kenya Kobayashi, Yuki Saito, Go Omura, Mizuo Ando, Takashi Sakamoto, Tatsuya Yamasoba, Takahiro Asakage
RESUMO
Head and neck squamous cell carcinoma of an unknown primary site (HNSCCUP) is a heterogeneous group of tumors that includes the human papillomavirus (HPV)-associated oropharyngeal squamous cell carcinoma. To investigate the relationship between HNSCCUP and HPV, we reviewed p16 overexpression and HPV DNA in lymph node metastases and examined their correlation with the primary site and clinical features. Thirty-three patients with HNSCCUP were retrospectively studied. Dissected neck metastases were analyzed for p16 overexpression by immunohistochemistry, and the presence of HPV DNA was investigated by in situ hybridization. Of the 33 patients, 8 (24%) exhibited p16 overexpression. p16-positive lymph node metastases contained significantly more HPV DNA and were most frequently associated with occult primary lesions in the oropharynx and a favorable prognosis. Patients with a lower alcohol consumption, only level II/III metastasis, and cystic lymph node metastasis tended to have p16 overexpression. This is the first report on the relationship of HNSCCUP with p16 and HPV DNA status in Asian patients. In total, 24% of the HNSCCUP patients were p16 positive. p16 overexpression in neck metastasis was predictive of both an occult primary lesion in the oropharynx and an association with HPV infection. Alcohol consumption, location, and features of neck metastasis were correlated with p16 expression.