Merck
  • Apelin inhibits the development of diabetic nephropathy by regulating histone acetylation in Akita mouse.
Apelin inhibits the development of diabetic nephropathy by regulating histone acetylation in Akita mouse.
The Journal of physiology (2013-11-20)
Hong Chen, Jianshuang Li, Lihua Jiao, Robert B Petersen, Jiong Li, Anlin Peng, Ling Zheng, Kun Huang
RESUMO
Diabetic nephropathy is the primary cause of end-stage renal disease. Increasing numbers of patients are suffering from this disease and therefore novel medications and therapeutic approaches are urgently needed. Here, we investigated whether apelin-13, the most active member of the adipokine apelin group, could effectively suppress the development of nephropathy in Akita mouse, a spontaneous type 1 diabetic model. Apelin-13 treatment decreased diabetes-induced glomerular filtration rate, proteinuria, glomerular hypertrophy, mesangial expansion and renal inflammation. The inflammatory factors, activation of NF-κB, histone acetylation and the enzymes involved in histone acetylation were further examined in diabetic kidneys and high glucose- or sodium butyrate-treated mesangial cells in the presence or absence of apelin-13. Apelin-13 treatment inhibited diabetes-, high glucose- and NaB-induced elevation of inflammatory factors, and histone hyperacetylation by upregulation of histone deacetylase 1. Furthermore, overexpression of apelin in mesangial cells induced histone deacetylation under high glucose condition. Thus, apelin-13 may be a novel therapeutic candidate for treatment of diabetic nephropathy via regulation of histone acetylation.