Merck
  • Epigenetic regulator MLL2 shows altered expression in cancer cell lines and tumors from human breast and colon.
Epigenetic regulator MLL2 shows altered expression in cancer cell lines and tumors from human breast and colon.
Cancer cell international (2010-05-04)
Thanemozhi G Natarajan, Bhaskar V Kallakury, Christine E Sheehan, Margaret B Bartlett, Natarajan Ganesan, Anju Preet, Jeffrey S Ross, Kevin T FitzGerald
RESUMO
MLL2, an epigenetic regulator in mammalian cells, mediates histone 3 lysine 4 tri-methylation (H3K4me3) through the formation of a multiprotein complex. MLL2 shares a high degree of structural similarity with MLL, which is frequently disrupted in leukemias via chromosomal translocations. However, this structural similarity is not accompanied by functional equivalence. In light of this difference, and previous reports on involvement of epigenetic regulators in malignancies, we investigated MLL2 expression in established cell lines from breast and colon tissues. We then investigated MLL2 in solid tumors of breast and colon by immunohistochemistry, and evaluated potential associations with established clinicopathologic variables. We examined MLL2 at both transcript and protein levels in established cell lines from breast and colon cancers. Examination of these cell lines showed elevated levels of MLL2. Furthermore, we also identified incomplete proteolytic cleavage of MLL2 in the highly invasive tumor cell lines. To corroborate these results, we studied tumor tissues from patients by immunohistochemistry. Patient samples also revealed increased levels of MLL2 protein in invasive carcinomas of the breast and colon. In breast, cytoplasmic MLL2 was significantly increased in tumor tissues compared to adjacent benign epithelium (p < 0.05), and in colon, both nuclear and cytoplasmic immunostaining was significantly increased in tumor tissues compared to adjacent benign mucosa (p < 0.05). Our study indicates that elevated levels of MLL2 in the breast and colon cells are associated with malignancy in these tissues, in contrast to MLL involvement in haematopoietic cancer. In addition, both abnormal cellular localization of MLL2 and incomplete proteolytic processing may be associated with tumor growth/progression in breast and colonic tissues. This involvement of MLL2 in malignancy may be another example of the role of epigenetic regulators in cancer.