Merck
  • Cytotoxic T Lymphocytes Regenerated from iPS Cells Have Therapeutic Efficacy in a Patient-Derived Xenograft Solid Tumor Model.
Cytotoxic T Lymphocytes Regenerated from iPS Cells Have Therapeutic Efficacy in a Patient-Derived Xenograft Solid Tumor Model.
iScience (2020-04-08)
Soki Kashima, Takuya Maeda, Kyoko Masuda, Seiji Nagano, Takamitsu Inoue, Masashi Takeda, Yuka Kono, Takashi Kobayashi, Shigeyoshi Saito, Takahiro Higuchi, Hiroshi Ichise, Yuka Kobayashi, Keiko Iwaisako, Koji Terada, Yasutoshi Agata, Kazuyuki Numakura, Mitsuru Saito, Shintaro Narita, Masaki Yasukawa, Osamu Ogawa, Tomonori Habuchi, Hiroshi Kawamoto
RESUMO
Current adoptive T cell therapies conducted in an autologous setting are costly, time consuming, and depend on the quality of the patient's T cells. To address these issues, we developed a strategy in which cytotoxic T lymphocytes (CTLs) are regenerated from iPSCs that were originally derived from T cells and succeeded in regenerating CTLs specific for the WT1 antigen, which exhibited therapeutic efficacy in a xenograft model of leukemia. In this study, we extended our strategy to solid tumors. The regenerated WT1-specific CTLs had a strong therapeutic effect in orthotopic xenograft model using a renal cell carcinoma (RCC) cell line. To make our method more generally applicable, we developed an allogeneic approach by transducing HLA-haplotype homozygous iPSCs with WT1-specific TCR α/β genes that had been tested clinically. The regenerated CTLs antigen-specifically suppressed tumor growth in a patient-derived xenograft model of RCC, demonstrating the feasibility of our strategy against solid tumors.