- Stabilisation of lipid membrane-incorporated porphyrin derivative aqueous solutions and their photodynamic activities.
Stabilisation of lipid membrane-incorporated porphyrin derivative aqueous solutions and their photodynamic activities.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2018-11-27)
Toshimi Nakaya, Banri Horiguchi, Shodai Hino, Kouta Sugikawa, Hisakage Funabashi, Akio Kuroda, Atsushi Ikeda
PMID30474674
RESUMO
Lipid membrane-incorporated porphyrin derivatives (LMIPors) having three phenyl moieties and one pyridyl or pyridinium moiety at the meso-positions were more stable than LMIPors having phenyl and/or pyridyl moieties. The former two LMIPors showed high photodynamic activity toward cancer cells under photoirradiation at wavelengths over 600 nm, which are the most suitable for photodynamic therapy. The photodynamic activities were greater than that of Photofrin, which is currently the main drug employed clinically as a photosensitiser. The results represent significant progress toward the optimisation of LMIPors as photosensitisers.