Merck
Rat monoclonal antibodies to mouse lung components for analysis of fibrosis.
Experimental and molecular pathology (1987-08-01)
S J Kennel, J A Hotchkiss, M C Rorvik, D P Allison, L J Foote
RESUMO
Six rat monoclonal antibodies to mouse lung membrane fraction have been characterized. Each has unique binding properties and can be used to stain particular lung components in paraffin sections. One antibody, 133-13A, recognizes a 100-kDa glycoprotein on lung tumor cells, but stains only macrophage-like cells in normal or fibrotic lung sections as determined by immunogold electron microscopy. The monoclonal antibody 273-34A binds to a 112-kDa protein on the surface of normal mouse lung fibroblasts. Immunogold electron microscopy demonstrates antibody binding to capillary endothelial cells, but not to fibroblasts. Type I, or Type II cells. Staining of fibrotic sections with MoAb 273-34A is dramatically enhanced over staining of normal lung. A third antibody, 370-8A, gives a general staining pattern throughout normal lung that is intensified in fibrotic lung. Another MoAb 328-41A mediates intense nuclear fluorescence of lung tumor cells and cells in lung sections. It binds to 14- and 17-kDa proteins that may be high mobility group (HMG) nuclear proteins. Enzyme inhibition studies and immunofluorescence staining patterns on normal lung indicate that elastin may be the target of MoAb 328-35B. MoAb 327-5B binds to normal mouse lung fibroblasts and red blood cell membranes. These last three MoAbs stain macrophages in fibrotic lung, but give a general pattern of light epithelial cell stain in normal lung sections indicating macrophage engulfment of the normal cell antigen during fibrosis. These antibodies should be useful in identifying cell types and molecular mechanisms involved in early stages of fibrosis induced by different chemical insults.