Merck
Zamak 410 - zinc/aluminum/copper/magnesium foam, thickness 50 mm, Zn95%/Al 4%/Cu 1%/Mg%, size 150 × 150 mm, bulk density 0.33 g/cm3
Zamak 410 - zinc/aluminum/copper/magnesium
Zamak 410 - zinc/aluminum/copper/magnesium foam, thickness 50 mm, Zn95%/Al 4%/Cu 1%/Mg%, size 150 × 150 mm, bulk density 0.33 g/cm3