Documentos Principais
Z180939
Ace auto-zero McLeod vacuum gauges
pressure 0-5 mmHg
Selecione um tamanho
Selecione um tamanho
Alterar visualização
About This Item
Código UNSPSC:
41112406
NACRES:
NB.79
fabricante/nome comercial
Ace Glass 872549
faixa de medição de pressão
0-5 mmHg
Categorias relacionadas
Descrição geral
A tilting gauge with a constant zero overflow which eliminates the capillary-depression error from an expanding meniscus. Once the zero line of the scale is aligned with the tip of the closed capillary, it is only necessary to bring the gauge to an upright position and read the scale behind the closed leg.
Gauges are fabricated from calibrated ACE Trubore tubing, to assure uniform and reproducible reading between the gauges. Mercury trap is incorporated in female joint, bulbs are enlarged. Joint is standard taper 20/65.
The pressure range is: 0-1.0 mmHg and the lowest reading is: 0.001 mmHg.
Mercury not included (approximately 20 mL of triple distilled mercury is needed to ffill this vacuum gauge to working volume).
Escolha uma das versões mais recentes:
Certificados de análise (COA)