Skip to Content
Merck
All Photos(2)

Key Documents

V900169

Sigma-Aldrich

Ammonium thiocyanate

Vetec, reagent grade, 98%

Synonym(s):

Ammonium rhodanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4SCN
CAS Number:
Molecular Weight:
76.12
Beilstein:
3595135
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
crystals

grade

reagent grade

vapor pressure

0.000114 hPa ( 20 °C)

product line

Vetec

Assay

98%

form

crystals

pH

4.5-6.0 (25 °C, 5% in water)

mp

152-154 °C (lit.)

SMILES string

N.SC#N

InChI

1S/CHNS.H3N/c2-1-3;/h3H;1H3

InChI key

SOIFLUNRINLCBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun Liu et al.
Human vaccines & immunotherapeutics, 16(7), 1699-1707 (2019-12-07)
Polysaccharides isolated from natural plants may represent a novel source of vaccine adjuvants. In this research, we focused on a natural plant polysaccharide, PCP-I, which is derived from Poria cocos, a Chinese traditional herbal medicine. We chose the anthrax protective
Xiaolian Gu et al.
The Journal of investigative dermatology, 135(8), 2077-2083 (2015-04-02)
Epigenetic modifications by DNA methylation are associated with a wide range of diseases. Previous studies in psoriasis have concentrated on epigenetic changes in immune cells or in total skin biopsies that include stromal-associated changes. In order to improve our understanding
Andrea Trost et al.
Experimental dermatology, 16(3), 185-190 (2007-02-09)
As global transcriptome analyses with a growing demand on layer-specific applications are widely used in cutaneous biology, we investigated the effect of established and optimized dermo-epidermal separation methods on the quality of RNA. We compared enzymatic separation with dispase, chemical
Leok Poh Chua et al.
Artificial organs, 32(5), 376-387 (2008-05-13)
The velocity fields within the impeller passages of three different impellers of the Kyoto-NTN bio-centrifugal ventricular assist device are measured using laser Doppler velocimetry in this study. The 16 forward-swept-blade impeller has better performance than the 16 straight-blade and 8
Mark A Zottola
Journal of molecular graphics & modelling, 28(2), 183-186 (2009-07-25)
Cyanide (CN) is considered to be a terrorist chemical weapon due to its ready availability in multi-kilogram quantities and multi-modal means of intoxication. The body uses the sulfur transferase enzyme rhodanese to detoxify cyanide via conversion of cyanide to thiocyanate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service