G005458
HPLC Analysis of mAb Charge Variants on TSKgel® CM-STAT
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: TSKgel CM-STAT, 10 cm x 4.6 mm I.D., 7 μm particles (821966)
mobile phase: [A] 20 mM MES buffer, pH 6.0; [B] 0.5 M NaCl in buffer A, pH 6.0
gradient: 0% B (0 min), 30% B (15 min), 100% B (15 min),
0% B (17 min), 10% B (17 min), 10% B (21 min)
flow rate: 1 mL/min
column temp.: ambient
detector: UV, 280 nm
injection: 20 μL
sample: monoclonal antibodies (mAb A through E)
suitability
application for HPLC
application(s)
life science and biopharma
Analysis Note
Citation
Legal Information
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service