27533
Vials, screw top with black polypropylene hole cap (10-425 thread), large opening, pre-assembled
volume 2 mL, clear glass vial, PTFE/silicone/PTFE septum, pkg of 100 ea
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
PTFE/silicone/PTFE septum
black polypropylene cap
clear glass vial
feature
closure type black polypropylene hole cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H × I.D.
12 mm × 32 mm × 6 mm
volume
2 mL
fitting
thread for 10-425
General description
Vials are suitable basically to inject samples from an autosampler. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of the sample from the vials. Screw top clear glass vials are used on many autosamplers with a large opening of 6 mm. The threads on the larger opening vials are 10-425. Includes a black polypropylene hole cap and a PTFE/silicone/PTFE septum, suitable for use with most organic solvents (until pierced) and not recommended for use with chlorosilanes.
Suitability
Autosampler compatibility:
PerkinElmer Autosystem
PE ISS-100, 200, Integral 4000 and Series 200
Shimadzu GC and LC Autosamplers
Spark Marathon, Midas, Triathlon
Thermo HPLC Spectra System
Waters Alliance
PerkinElmer Autosystem
PE ISS-100, 200, Integral 4000 and Series 200
Shimadzu GC and LC Autosamplers
Spark Marathon, Midas, Triathlon
Thermo HPLC Spectra System
Waters Alliance
Other Notes
The diameter of inserts varies between manufacturers. Only the following insert part numbers are compatible with these large opening vials.
- 24719, 24720Glass insert, conical bottom, 6 x 29mm
- 24721Glass insert with bottom spring, 6 x 29mm
- 24722Polypropylene insert with bottom spring, 6 x 29mm
- 24717, 24718Glass insert, conical bottom, 6 x 31mm
- 24715, 24716Glass insert, flat bottom, 6 x 31mm
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)