Key Documents
Z659452
Bandelin SONOREX PR 140 C pipette washer
supplied with basket, lid, cleaning concentrate TICKOPUR R 33 - 5 litres
About This Item
UNSPSC Code:
41103706
AC/DC input
230 V AC, 50 - 60 Hz
feature
CE compliant
supplied with basket, lid, cleaning concentrate TICKOPUR R 33 - 5 litres
manufacturer/tradename
Bandelin
availability
not available in USA and Canada
General description
Fast cleaning. No time-consuming washing. Rinsing process in the same vessel according to the siphon principle - no shifting around. Accelerated circulation of pipettes. No broken glass when used according to the operating instructions. Also suitable for pipettes of plastic, burettes and other glassware.
Max. length of objects to be cleaned765 mm
Operating capacity13.9 L
Height of the device1130 mm
Required floor space335 × 255 mm
HF-power2 × 450 W, 35 kHz
Radiating surface diameter150 mm
Timer1 to 60 min.
RFI-proof and CE marked
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)