Skip to Content
Merck
All Photos(1)

Key Documents

Z334014

BRAND® microcentrifuge tube, 1.5 mL with lid, PP

yellow, pack of 500

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

material

cap (Lid)
polypropylene

sterility

non-sterile

feature

RCF 20,000 × g

packaging

pack of 500

manufacturer/tradename

BRAND

H × O.D.

41 mm × 10.75 mm

capacity

1.5 mL

membrane diam. × thickness

8.5 mm × 0.3 mm

color

yellow

Looking for similar products? Visit Product Comparison Guide

General description

Uniform lid membrane thickness ensures trouble-free piercing by the analyser. Lid provides leak-free seal and fits tightly, yet is easily reopened. Matted labeling area with subdivisions for approximate volume determination. Withstands RCF 20000 × g at 20°C for 20 minutes.

* The amber-colored tubes are particularly suitable for light sensitive reagents.

Other Notes

BR780521 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service