SRP8036
IL-36β mouse
>95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized
Synonym(s):
Interleukin-1 family member 8, Interleukin-36β, Sentrin-specific protease 2
About This Item
Product Name
IL-36β mouse, recombinant, expressed in E. coli, untagged, >95% (SDS-PAGE)
biological source
mouse
recombinant
expressed in E. coli
conjugate
untagged
Assay
>95% (SDS-PAGE)
form
lyophilized
mol wt
~17 kDa
packaging
pkg of 10 μg
impurities
<0.01 EU/μg endotoxin, tested
color
white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Il36b(69677)
General description
Physical form
Reconstitution
Other Notes
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service