Skip to Content
Merck
All Photos(1)

Key Documents

SRP2052

Sigma-Aldrich

p75-CTR (C-terminal region) human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

DFS70, LEDGF, MGC74712, PAIP, PSIP2, p52, p75

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~24.4 kDa

packaging

pkg of 10 μg

concentration

600 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PSIP1(11168)

General description

p75 was first identified during micro-sequencing of a protein that co-purified with positive cofactor 4 (PC4), a general transcriptional coactivator. This is a transcriptional coactivator and has a molecular weight of 75kDa. This protein is also known as LEDGF (lens epithelium-derived growth factor), and belongs to the hepatoma-derived growth factor (HDGF) family. p75 is constitutively expressed in the nucleus. This gene is localized to human chromosome 9p22.3, and is composed of 15 exons and 14 introns. It is transcribed into two splice variants- p52 and p75 both of which contain 325 residues in their N-terminals. They differ in their C-terminal, where p75 contains 205 residues in its C-terminal. Its C-terminal also contains the 83-residue IN binding domain (IBD) from residues 340–417, which is essential for p75′s virological and cellular characteristics.

Biochem/physiol Actions

Lens epithelium-derived growth factor (LEDGF, also known as p75) has been shown to enhance survival of lens epithelial cells (LECs) against stress. LEDGF is a transcriptional activator. It protects the cells by binding to cis-stress response ((A/T)GGGG(T/A)), and heat shock (HSE; nGAAn) elements in the stress genes and activating their transcription. Originally, it was isolated as a co-activator required for transcriptional activation in human cell-free systems containing RNA polymerase II and general initiation factors. LEDGF is expressed at all stages of development in a variety of organs and tissues. A second protein product, p52, can be produced from the same gene due to alternative splicing of pre-mRNA. in vitro, p52 was found to be more general and stronger transcriptional co-activator than LEDGF/p75. HIV-1 integrase (IN) forms a specific nuclear complex with p75 but not with p52, suggesting a role for p75′s C-terminal region in retroviral integration.
p75 and p52 proteins associate with chromatin tightly and interact with other nuclear proteins, out of which multiple proteins are anchored by p75 to the chromatin fiber. Therefore, these proteins are involved in transcriptional regulation. Lentiviruses utilize the chromatin tethering capacity of this protein to integrate into the host chromosomes. p75 not only anchors HIV (human immunodeficiency virus) integrase to chromatin and prevents its degradation, but also promotes the genome-wide HIV integration. Human papillomavirus (HPV) E6/E7 oncogenes promote the expression of p75 in host cells, which in turn confers protection to HPV-positive cancer cells against different types of cellular stress, such as DNA damage.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oncogenic human papillomaviruses activate the tumor-associated lens epithelial-derived growth factor (LEDGF) gene.
Leitz J et al
PLoS Pathogens, 10(3), e1003957-e1003957 (2014)
D P Singh et al.
Biochemical and biophysical research communications, 283(4), 943-955 (2001-05-15)
We have investigated the mechanism by which LEDGF protects cells against environmental stress. Our earlier report showed that a low level of LEDGF was present in the nucleus of most cell types and significant elevation of LEDGF level was induced
Integrase, LEDGF/p75 and HIV replication.
Poeschla EM
Cellular and Molecular Life Sciences, 65(9), 1403-1424 (2008)
N Fatma et al.
The Journal of biological chemistry, 276(52), 48899-48907 (2001-10-26)
Antioxidant protein 2 (AOP2), a member of the newly defined family of thiol-specific antioxidant proteins, has been shown to remove H(2)O(2) and protect proteins and DNA from oxidative stress. Here we report that LEDGF is one of the regulatory factors
Virological and cellular roles of the transcriptional coactivator LEDGF/p75.
Llano M et al
Current Topics in Microbiological Immunology, 339, 125-146 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service