Skip to Content
Merck
All Photos(1)

Key Documents

SCP0219

Sigma-Aldrich

ProAM N20 Peptide

≥95% (HPLC), lyophilized, human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C112H178N36O27
Molecular Weight:
2460.84
UNSPSC Code:
12352202
NACRES:
NA.32

product name

ProAM N20 human,

biological source

human

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ala-Arg-Leu-Asp-Val-Ala-Ser-Glu-Phe-Arg-Lys-Lys-Trp-Asn-Lys-Trp-Ala-Leu-Ser-Arg-NH2

Application

Proadrenomedullin N-terminal 20 peptide (PAMP, ProAM N20) is a novel peptide found in adrenomedullin precursor that may be involved in hypotension and the control of reproductive physiology.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service