SAB5200257
Monoclonal Anti-Vacht - Streptavidin antibody produced in mouse
clone S6-38, purified immunoglobulin
Synonym(s):
Anti-MGC12716, Anti-Slc18a3, Anti-Vesicular Acetylcholine Transporter
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
streptavidin conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
S6-38, monoclonal
form
buffered aqueous solution
species reactivity
human, mouse, rat
concentration
1 mg/mL
technique(s)
immunohistochemistry: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... Slc18A3(6572)
Specificity
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
SKU | GTIN |
---|---|
SAB5200257-100UG | 4061832155425 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service