Skip to Content
Merck
All Photos(2)

Key Documents

SAB2702179

Sigma-Aldrich

Anti-ERCC3 antibody produced in rabbit

Synonym(s):

GTF2H, RAD25, TFIIH, XPB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: 1:500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ERCC3(2071)

Immunogen

Peptide corresponding to the carboxy-terminus of human XPB.

Application

For WB: Use at a dilution of 1:500 and incubate at RT for 40 minutes. Blocking solution: 3% Milk/TBST at RT for one hour. Optimal dilutions/concentrations should be determined by the researcher.

Biochem/physiol Actions

The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne′s syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, pH 7.2, containing no preservatives

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service