Skip to Content
Merck
All Photos(2)

Key Documents

SAB2100031

Sigma-Aldrich

Anti-ACP2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Acid phosphatase 2, lysosomal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
horse, human, mouse, dog, rat
citations:
technique(s):
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

horse, human, mouse, dog, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACP2(53)

Immunogen

Synthetic peptide directed towards the middle region of human ACP2

Biochem/physiol Actions

ACP2 is the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Lysosomal acid phosphatase is comprised of two subunits, alpha and beta, and is chemically and genetically distinct from red cell acid phosphatase. Lysosomal acid phosphatase 2 is a member of a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Acid phosphatase deficiency is caused by mutations in the ACP2 (beta subunit) and ACP3 (alpha subunit) genes. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: VPITEDRLLKFPLGPCPRYEQLQNETRQTPEYQNESSRNAQFLDMVANET

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service