Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400176

Sigma-Aldrich

Anti-NAP1L2 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-BPX, Anti-MGC26243

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NAP1L2(4674)

General description

This gene encodes a member of the nucleosome assembly protein (NAP) family. The function of this family member is unknown; however, mouse studies suggest that it represents a class of tissue-specific factors interacting with chromatin to regulate neuronal cell proliferation. (provided by RefSeq)

Immunogen

NAP1L2 (AAH26325.1, 1 a.a. ~ 460 a.a) full-length human protein.

Sequence
MAKSENRKELSESSQEEAGNQIMVEGLGEHLERGEDAAAGLGDDGKCGEEAAAGLGEEGENGEDTAAGSGEDGKKGGDTDEDSEADRPKGLIGYVLDTDFVESLPVKVKYRVLALKKLQTRAANLESKFLREFHDIERKFAEMYQPLLEKRRQIINAIYEPTEEECEYKSDSEDCDDEEMCHEEMYGNEEGMVHEYVDEDDGYEDYYYDYAVEEEEEEEEEDDIEATGEENKEEEDPKGIPDFWLTVLKNVDTLTPLIKKYDEPILKLLTDIKVKLSDPGEPLSFTLEFHFKPNEYFKNELLTKTYVLKSKLAYYDPHPYRGTAIEYSTGCEIDWNEGKNVTLKTIKKKQKHRIWGTIRTVTEDFPKDSFFNFFSPHGITSNGRDGNDDFLLGHNLRTYIIPRSVLFFSGDALESQQEGVVREVNDAIYDKIIYDNWMAAIEEVKACCKNLEALVEDIDR

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service