RAB0338
Mouse Growth Arrest Specific 6 ELISA Kit
for serum, plasma and cell culture supernatant
Synonym(s):
Gas6
About This Item
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type plasma
sample type cell culture supernatant(s)
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 20.48-5000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Lgals7(16858)
General description
Immunogen
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
- RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS
- RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS
- RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS
- RABSTOP3ELISA Stop Solution (Item I)SDS
- RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS
- RABWASH420X Wash Buffer (Item B)SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service