Skip to Content
Merck
All Photos(2)

Key Documents

MAK213

Sigma-Aldrich

Neutrophil Elastase Inhibitor Screening Kit

Sufficient for 100 Fluorometric tests

Synonym(s):

Neutrophil Elastase Inhibition Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200
NACRES:
NA.25

detection method

fluorometric

storage temp.

−20°C

Gene Information

human ... ELANE(1991)
mouse ... ELANE(50701)
rat ... ELANE(299606)

General description

Neutrophil elastase (NE) is a serine protease that hydrolyzes proteins within the azurophilic granules of the neutrophils. Once secreted, it digests collagen-VI and elastin of the extracellular matrix. Neutrophil elastase is implicated in cystic fibrosis, bronchiectasis, chronic obstructive pulmonary disease, congenital neutropenia, and lung cancer.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the screening of inhibitors of Neutrophil Elastase.

Principle

The Neutrophil Elastase Inhibitor Screening Kit is a rapid, sensitive and high throughput assay to screen and characterize potential inhibitors of NE. NE activity is measured by hydrolyzing the substrate to yield a fluorescent product (λex = 400/λem = 505 nm) proportional to the enzymatic activity present.

Kit Components Only

Product No.
Description

  • Assay Buffer

  • Substrate

  • Neutrophil Elastase

  • Inhibitor Control, 3 mM SPCK

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

ELANE mutations in cyclic and severe congenital neutropenia: genetics and pathophysiology.
Horwitz M S, et al.
Hematology/oncology Clinics of North America, 27, 19?41-19?41 (2013)
SVSDVSSDVD
PESTICIDES null
Neutrophil elastase as a target in lung cancer.
Moroy G, et al.
Anticancer Agents in Medicinal Chemistry, 12, 565?579-565?579 (2012)
Neutrophil elastase inhibitors: recent advances in the development of mechanism-based and nonelectrophilic inhibitors.
Sjo P, et al.
Future medicinal chemistry, 4(5), 651-660 (2012)
DWQDWQCW
FWQFQWFW
13th ACS Winter Fluorine Conference, paper No. 8 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service