Skip to Content
Merck
All Photos(1)

Key Documents

M1289

Sigma-Aldrich

Mounting Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

form

viscous liquid

technique(s)

immunocytochemistry: suitable

Application

For use in Mycoplasma stain kit.

Components

Contains sodium phosphate and citric acid in glycerol.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanbyeol Lee et al.
Toxicology letters, 240(1), 140-148 (2015-11-08)
Cigarette smoking is the major aetiologic factor in chronic obstructive pulmonary disease (COPD). Lung fibroblasts are key participants in the maintenance of the extracellular matrix within the lung parenchyma. However, it still remains unknown how pulmonary fibroblasts are affected by
Brigitte Grosse et al.
Tissue barriers, 1(4), e25210-e25210 (2014-03-26)
Tight junctions (TJs) of cells expressing simple epithelial polarity have been extensively studied, but less is known about TJs of cells expressing complex polarity. In this paper we analyzed, TJs of four different lines, that form bile canaliculi (BC) and
Jooyeon Lee et al.
In vitro cellular & developmental biology. Animal, 54(7), 505-512 (2018-06-06)
L-carnosine, a dipeptide of the amino acids β-alanine and histidine, is found in various tissues, such as the central nervous system and skeletal muscles. Recently, L-carnosine has been reported to possess anti-tumor activity; however, the molecular mechanism underlying its activity
Lorenz Adlung et al.
Cell reports, 36(6), 109507-109507 (2021-08-12)
Survival or apoptosis is a binary decision in individual cells. However, at the cell-population level, a graded increase in survival of colony-forming unit-erythroid (CFU-E) cells is observed upon stimulation with erythropoietin (Epo). To identify components of Janus kinase 2/signal transducer
Michael X Le et al.
Scientific reports, 6, 37215-37215 (2016-11-18)
Class switch recombination (CSR) in B cells requires the timely repair of DNA double-stranded breaks (DSBs) that result from lesions produced by activation-induced cytidine deaminase (AID). Through a genome-wide RNAi screen, we identified Kin17 as a gene potentially involved in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service