Skip to Content
Merck
All Photos(1)

Key Documents

M01070-HH

Sigma-Aldrich

Phosphate-ON Phosphoramidite

Synonym(s):

Phosphate-ON amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H62N7O11P
Molecular Weight:
956.03
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OC(=O)NCCCNC(=O)C[C@@H](CC#N)OP(OCCC#N)N(C(C)C)C(C)C)N3C=C(C)C(=O)NC3=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C49H62N7O11P/c1-33(2)56(34(3)4)68(64-28-11-24-50)67-41(23-25-51)29-44(57)52-26-12-27-53-48(60)66-42-30-45(55-31-35(5)46(58)54-47(55)59)65-43(42)32-63-49(36-13-9-8-10-14-36,37-15-19-39(61-6)20-16-37)38-17-21-40(62-7)22-18-38/h8-10,13-22,31,33-34,41-43,45H,11-12,23,26-30,32H2,1-7H3,(H,52,57)(H,53,60)(H,54,58,59)/t41-,42+,43-,45-,68?/m1/s1

InChI key

MCGIOYZUTKDSIL-NNUNARBDSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

This product is covered under US Patent Number 7,276,598

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service