Skip to Content
Merck
All Photos(2)

Key Documents

HPA031044

Sigma-Aldrich

Anti-INPP5K antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SKIP, Anti-inositol polyphosphate-5-phosphatase K

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL

immunogen sequence

LVFAKYQHLPYIQILSTKSTPTGLFGYWGNKGGVNICLKLYGYYVSIINCHLPPHISNNYQRLEHFDRILEMQNCEGRDIPNILDHDLIIWFGDMNFRIEDFGLHFVRESIKNRCYGGLWEKDQLSI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... INPP5K(51763)

General description

The INPP5K (inositol polyphosphate-5-phosphatase K) gene is mapped to human chromosome 17p13.3 The gene encodes for inositol polyphosphate-5-phosphatase K, also known as SKIP (skeletal muscle and kidney enriched inositol phosphatase). The protein is widely expressed in the brain and muscle. INPP5K localizes to endoplasmic reticulum and to actin ruffles in the cytoplasm.

Immunogen

inositol polyphosphate-5-phosphatase K recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

INPP5K (inositol polyphosphate-5-phosphatase K) is known to associate with insulin signaling in skeletal muscle by interacting with Pak1 (p21-activated kinase 1) and glucose-regulated protein 78. INPP5K suppresses insulin-dependent phosphatidylinositol 3,4,5-triphosphate generation, glucose incorporation, and membrane ruffle formation in skeletal muscle. INPP5K plays an important role in muscle cell differentiation and requires a muscle-specific transcription factor MyoD, for its expression. Mutations in INPP5K might cause congenital muscular dystrophy syndrome characterized with short stature, cataracts, and intellectual disability. Overexpression of INPP5K suppresses HBV (hepatitis B virus) gene expression and replication in hepatocellular carcinoma cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71216

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucose-regulated protein 78 (GRP78) binds directly to PIP3 phosphatase SKIP and determines its localization.
Ijuin T
Genes Cells, 21(5), 457-465 (2016)
Mutations in INPP5K Cause a Form of Congenital Muscular Dystrophy Overlapping Marinesco-Sjogren Syndrome and Dystroglycanopathy.
Osborn DPS
American Journal of Human Genetics, 100(3), 537-545 (2017)
Suppression of hepatitis B viral gene expression by phosphoinositide 5-phosphatase SKIP.
Hung CS
Cellular Microbiology, 11(1), 37-50 (2009)
Regulation of insulin signaling by the phosphatidylinositol 3,4,5-triphosphate phosphatase SKIP through the scaffolding function of Pak1.
Ijuin T and Takenawa T
Molecular and Cellular Biology, 32(17), 3570-3584 (2012)
Takeshi Ijuin et al.
The Journal of biological chemistry, 287(37), 31330-31341 (2012-07-21)
Insulin-like growth factors (IGFs) are essential for the development, regeneration, and hypertrophy of skeletal muscles. IGF-II promotes myoblast differentiation through phosphatidylinositol 3-kinase (PI 3-kinase), Akt, and mTOR signaling. Here, we report that skeletal muscle- and kidney-enriched inositol polyphosphate phosphatase (SKIP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service