Skip to Content
Merck
All Photos(1)

Key Documents

NO36 cell line human

10092312, human lung, bronchoalveolar, epithelial-like. Thick stellate cells with vacuoles.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human lung (bronchoalveolar)
growth mode:
Adherent
karyotype:
Not specified
morphology:
Epithelial-like: Thick stellate shaped cells with vacuoles.
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

NO36 cell line human, 10092312

biological source

human lung (bronchoalveolar)

description

Human malignant mesothelioma

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like: Thick stellate shaped cells with vacuoles.

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human malignant mesothelioma

Cell Line Description

This cell line was derived from the pleural fluid of a patient with malignant mesothelioma. The patient had known exposure to crocidolite asbestos. NO36 is not tumourigenic in nude mice. Cells express cytokeratin and epithelial membrane antigen (EMA) but not mucin. Cells are thick and stellate shaped with vacuoles. Cell Bank Australia reference number: CBA0142

Application

Investigation of the properties of malignant mesothelioma cells and the development of new diagnostic methods.

Culture Medium

RPMI 1640 (with 2mM L-Glutamine+25mM HEPES) + 5% FCS

Subculture Routine

Optimal split ratio 1:8 (seeding density 104 cells/cm2).Cells incubated at 37°C with 5% CO2.Harvest the cells using 0.05% Trypsin/EDTA at 37°C for 5 min.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Richard Lake - University of Western Australia, Australia

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service