C4985
Cytochrome P450 human
4F2 isozyme microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
form
solution
mol wt
45-60 kDa
packaging
vial of 0.5 nmol
UniProt accession no.
application(s)
cell analysis
storage temp.
−70°C
Gene Information
human ... CYP4F2(8529)
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules, both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds (drugs or toxins). Some isoforms have narrow substrate specificity, while others are promiscuous. The CYP1A1 isoform catalyzes 7-deethylation of ethoxyresorufin. Cytochrome P450 (CYP) plays an important role in detoxifying xenobiotics, cellular metabolism and homeostasis. One of the main mechanisms of drug-drug interactions is the induction or inhibition of these enzymes. CYP enzymes are transcriptionally activated by a variety of xenobiotics and by endogenous substrates via receptor-dependent pathways. Inhibition of these enzymes is a major factor in metabolism-based drug-drug interactions, and many chemotherapeutic medications can cause drug interactions by either inhibiting or inducing the cytochrome p450 enzyme system.
The CYP4F2 isoform catalyzes the 20-hydroxylation of leukotriene B4.
Unit Definition
One unit will hydroxylate 1 picomole of leukotriene B4 per minute at pH 7.4 at 37°C.
Physical form
Solution in 100 mM potassium phosphate buffer, pH 7.4.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of biological chemistry, 274(30), 21191-21199 (1999-07-20)
Cytochrome P450 4F3 (CYP4F3) catalyzes the inactivation of leukotriene B(4) by omega-oxidation in human neutrophils. To understand the regulation of CYP4F3 expression, we analyzed the CYP4F3 gene and cloned a novel isoform (CYP4F3B) that is expressed in fetal and adult
PloS one, 9(4), e94962-e94962 (2014-04-17)
The present study characterized in vitro metabolites of 20(R)-25-methoxyl-dammarane-3β, 12β, 20-triol (20(R)-25-OCH3-PPD) in mouse, rat, dog, monkey and human liver microsomes. 20(R)-25-OCH3-PPD was incubated with liver microsomes in the presence of NADPH. The reaction mixtures and the metabolites were identified
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1438-1446 (2014-07-06)
Cilengitide is a stable cyclic pentapeptide containing an Arg-Gly-Asp motif responsible for selective binding to αVβ3 and αVβ5 integrins. The candidate drug showed unexpected inhibition of cytochrome P450 (P450) 3A4 at high concentrations, that is, a 15-mM concentration caused attenuation
Drug metabolism and disposition: the biological fate of chemicals, 42(10), 1727-1736 (2014-07-16)
Diosbulbin B (DIOB), a furan-containing diterpenoid lactone, is the most abundant component of Dioscorea bulbifera L. (DB), a traditional Chinese medicine herb. Administration of purified DIOB or DB extracts has been reported to cause liver injury in animals. The mechanisms
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service