Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR24C

Corning® Axygen® 24 Well PCR Microplate

clear, size 24 wells, : no, conical bottom clear polypropylene wells, maximum volume 200 μL, pkg of 5x10plates/cs

Synonym(s):

Corning® Axygen® PCR Microplate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

conical bottom clear polypropylene wells

sterility

non-sterile

feature

: no

manufacturer/tradename

Axygen Inc PCR-24-C
Corning

packaging

pkg of 5x10plates/cs

maximum volume

200 μL

size

24 wells

working volume

200 μL

color

clear

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service