Skip to Content
Merck
All Photos(1)

Key Documents

APREST95500

Sigma-Aldrich

PrEST Antigen MAP4K5

Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous solution

Synonym(s):

GCKR, KHS, KHS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

recombinant

expressed in E. coli

product line

Prestige Antibodies® Powered by Atlas Antibodies

Assay

>80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

predicted mol wt 23 kDa

purified by

immobilized metal affinity chromatography (IMAC)

concentration

≥0.5 mg/mL

sequence note

WLYVINNTLMSLSEGKTFQLYSHNLIALFEHAKKPGLAAHIQTHRFPDRI

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAP4K5(11183)

General description

N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G)

Application

Suitable as a blocking agent and positive assay control using corresponding antibodies.

Linkage

Corresponding Antibodies HPA078030

Physical form

Solution in 1 M urea-PBS, pH 7.4

Storage and Stability

Store at -20°C. Avoid repeated freeze/thaw cycles.

Other Notes

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service