Skip to Content
Merck
All Photos(1)

Key Documents

33392

Supelco

Chloramben

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H2(Cl)2CO2H
CAS Number:
Molecular Weight:
206.03
Beilstein:
2365906
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

mp

194-197 °C (dec.) (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1cc(Cl)cc(C(O)=O)c1Cl

InChI

1S/C7H5Cl2NO2/c8-3-1-4(7(11)12)6(9)5(10)2-3/h1-2H,10H2,(H,11,12)

InChI key

HSSBORCLYSCBJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloramben is a chlorinated, ammonium or sodium salt based, pre-emergence herbicide used to control a broad spectrum of weeds in vegetable and field crops. Its mode of action involves the inhibition of seedling root development in plants.

Application

Chloramben may be used as an analytical reference standard for the determination of the analyte in water samples using liquid chromatography with electrochemical detection (LC-EC).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agrochemical and Pesticide Desk Reference on CD-ROM (1999)
Liquid chromatographic determination of selected carbamate pesticides in water with electrochemical detection.
Anderson LJ and Chesney JD
Analytical Chemistry, 52(13), 2156-2161 (1980)
Plant Growth Substances (1996)
[Establishment of the maximum permissible concentration of amiben in the water of reservoirs].
S Iu Buslovich et al.
Gigiena i sanitariia, (4)(4), 74-75 (1980-04-01)
Field persistence studies with triallate and trifluralin both singly and in combination with chloramben.
A E Smith et al.
Bulletin of environmental contamination and toxicology, 29(2), 240-242 (1982-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service