Skip to Content
Merck
All Photos(1)

Key Documents

21177

Sigma-Aldrich

Calcium phosphate dibasic

puriss. p.a., anhydrous, >98.5% (KT)

Synonym(s):

Calcium hydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaHPO4
CAS Number:
Molecular Weight:
136.06
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

puriss. p.a.

Assay

>98.5% (KT)

form

powder

impurities

≤0.2% insoluble matter in HCl

loss

6.5-8.5% loss on ignition, 850 °C

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤1000 mg/kg

cation traces

As: ≤0.5 mg/kg
Ba: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤200 mg/kg
K: ≤50 mg/kg
Mg: ≤2000 mg/kg
Mn: ≤100 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤200 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Ca++].OP([O-])([O-])=O

InChI

1S/Ca.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+2;/p-2

InChI key

FUFJGUQYACFECW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria Pauli et al.
Journal of pharmaceutical sciences, 108(6), 2041-2055 (2019-01-25)
In line with the ongoing shift from batch to continuous pharmaceutical production of solid oral dosage forms, a novel continuous fluid-bed dryer was developed. The forced feed nature of the Glatt GPCG2 CM fluid-bed dryer allows continuous, first-in-first-out drying of
Fernando Notario-Pérez et al.
Carbohydrate polymers, 179, 305-316 (2017-11-08)
The use of sustained-release mucoadhesive vaginal tablets of antiretroviral drugs as microbicidal formulations can be an effective strategy for reducing the sexual transmission of HIV from men to women, which is a main problem particularly in low- and middle-income countries.
Fernando Notario-Pérez et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 117, 204-215 (2018-02-27)
Sustained-release vaginal microbicides hold out great hope for the prevention of sexual transmission of HIV from men to women. Tenofovir (TFV) -an antiretroviral drug- sustained-release vaginal compacts combining two release control systems (by drug-loading granules with hydrophobic polymers and incorporating
Michaela Rödel et al.
Journal of tissue engineering, 9, 2041731418788509-2041731418788509 (2018-07-27)
Silk fibroin is commonly used as scaffold material for tissue engineering applications. In combination with a mineralization with different calcium phosphate phases, it can also be applied as material for bone regeneration. Here, we present a study which was performed
Victoria Pauli et al.
Journal of pharmaceutical and biomedical analysis, 162, 101-111 (2018-09-19)
The use of Near Infrared Spectroscopy (NIRS) as a fast and non-destructive technique was employed for the control and monitoring of the tableting step during a continuous manufacturing process. Two NIRS methods were optimized in order to in-line control the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service