Skip to Content
Merck
All Photos(1)

Key Documents

14849

Supelco

Triethyl citrate

analytical standard

Synonym(s):

Ethyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOC2H5)(CH2COOC2H5)2
CAS Number:
Molecular Weight:
276.28
Beilstein:
1801199
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E1505
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

9.7 (vs air)

vapor pressure

1 mmHg ( 107 °C)

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.442 (lit.)
n20/D 1.443

bp

235 °C/150 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

InChI

1S/C12H20O7/c1-4-17-9(13)7-12(16,11(15)19-6-3)8-10(14)18-5-2/h16H,4-8H2,1-3H3

InChI key

DOOTYTYQINUNNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.
Triethyl citrate is an ester of citric acid, commonly used as a sequestrant in beverages. It is also reported as an effective plasticizing agent.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Triethyl citrate may be used as an analytical standard for the quantification of the analyte in pharmaceutical formulations using high-performance liquid chromatography (HPLC) with UV detection and leaves of Coriandrum sativum and Melia Azedarach medicinal plants using gas chromatography-mass spectrometry (GC-MS).

Other Notes

Ion Mobility: TWCCSN2 value of 165.5 Å2 [M+Na]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 14849 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dictionary of Food ingredients (2013)
Development and validation of a gradient HPLC method for the determination of clindamycin and related compounds in a novel tablet formulation.
Platzer DJ, White BA.
Journal of Pharmaceutical and Biomedical Analysis, 41(1), 84-88 (2006)
M R Abbaspour et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 68(3), 747-759 (2007-11-06)
One challenge in tableting of sustained-release multiparticulates is maintaining the desired drug release after compaction. The aim of this study was to design sustained-release ibuprofen tablets which upon oral ingestion rapidly disintegrate into sustained-release pellets in which the integrity of
Soravoot Rujivipat et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 81(1), 223-229 (2012-01-25)
Enteric polymers such as cellulose esters (cellulose acetate phthalate, hydroxypropylmethylcellulose acetate succinate) and methacrylic acid-acrylate copolymers (Eudragit® L100-55 and S100) are quite brittle in the dry state and thus not suitable as pellet coatings for compression into tablets. The objective
Kakuyou Ogawa et al.
Biological & pharmaceutical bulletin, 37(9), 1559-1563 (2014-09-02)
Zerumbone 1 is an 11-membered cyclic sesquiterpene obtained from the rhizomes of Zingiber zerumbet SMITH (Zingiberaceae). In this study, we investigated the structure-activity relationship of 1, α-humulene (2), tetrahydrozerumbone stereoisomers (3-5), and tetrahydrozerumbone derivatives (6-9). The oxygen-containing functional groups and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service