01-4530
Ammonium carbonate
SAJ special grade, ≥30.0% NH3 basis
Synonym(s):
Hartshorn salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(NH4)2CO3
CAS Number:
Molecular Weight:
96.09
Beilstein:
4253487
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
Recommended Products
grade
SAJ special grade
vapor density
2.7 (vs air)
Assay
≥30.0% NH3 basis
form
solid
availability
available only in Japan
storage temp.
15-25°C
SMILES string
N.N.OC(O)=O
InChI
1S/CH2O3.2H3N/c2-1(3)4;;/h(H2,2,3,4);2*1H3
InChI key
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
N N Mal'tseva et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 62(3), 56-65 (2000-08-10)
The intensification of activity of associative and symbiotic nitrogen fixation while using low amounts of ammonium-carbonate compounds, carbonic-ammonium salts, in particular, has been established. The main ways and mechanisms of the action of ammonium-carbonate salts as a new and perspective
Debabrata Nandi et al.
Journal of colloid and interface science, 454, 69-79 (2015-05-26)
Reduced-graphene-oxide based superparamagnetic nanocomposite (GC) was fabricated and applied for the remediation of Ni(II) from an aqueous medium. The as-prepared GC was extensively characterized by Raman, TEM, AFM, SEM-EDX, SQUID, and BET analyses. Quantitative immobilization of Ni(II) in an aqueous
Tomohisa Hasunuma et al.
Biotechnology for biofuels, 7(1), 493-493 (2015-02-05)
To ensure reliable sources of energy and raw materials, the utilization of sustainable biomass has considerable advantages over petroleum-based energy sources. Photosynthetic algae have attracted attention as a third-generation feedstock for biofuel production, because algae cultivation does not directly compete
Itsuo Murakami et al.
PloS one, 9(12), e115359-e115359 (2014-12-17)
Resveratrol (RESV) is a plant polyphenol, which is thought to have beneficial metabolic effects in laboratory animals as well as in humans. Following oral administration, RESV is immediately catabolized, resulting in low bioavailability. This study compared RESV metabolites and their
Mirosława Różycka et al.
PloS one, 9(12), e114308-e114308 (2014-12-10)
Fish otoliths, biominerals composed of calcium carbonate with a small amount of organic matrix, are involved in the functioning of the inner ear. Starmaker (Stm) from zebrafish (Danio rerio) was the first protein found to be capable of controlling the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service