MABS2287
Anti-Jagged2 Antibody, clone HMJ2-1

Synonym(s):
D12Ggc2e, JAG2, Protein jagged-2, mJagged2-1, syndactylism
About This Item
biological source
hamster
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
HMJ2-1, monoclonal
mol wt
calculated mol wt 134.75 kDa
purified by
using protein G
species reactivity
mouse
packaging
antibody small pack of 100
technique(s)
ELISA: suitable
flow cytometry: suitable
inhibition assay: suitable
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
storage temp.
-10 to -25°C
Gene Information
mouse ... Jag2(16450)
Specificity
Immunogen
Application
Evaluated by Flow Cytometry in CHO cells transfected with mouse Jagged2.
Flow Cytometry Analysis: 1.0 g of this antibody detected Jagged2 in one million CHO cells transfected with mouse Jagged2.
Tested Applications
Flow Cytometry Analysis: A 1:500 dilution from a representative lot detected Jagged2 in CHO cells transfected with mouse Jagged2 (Courtesy of Dr. Hideo Yagita, Ph.D., Juntendo University, Japan).
ELISA: A representative lot detected Jagged2 in ELISA application (Sekine, C., et al. (2014). Arthritis Rheumatol. 66(10):2751-61).
Flow Cytometry Analysis: A representative lot detected Jagged2 in Flow Cytometry application (Kijima, M., et al. (2008). Proc Natl Acad Sci USA. 105(19):7010-5).
Agonist: A representative lot promoted Treg cell differentiation and enhanced Jagged-2 Notch signaling via activation of RBP-Jk (canonical Notch pathway). (Elyaman, W., et al. (2012). Immunity. 36(4):623-34; Riella, L.V., et al. (2013). Eur J Immunol. 43(6):1449-58).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service