Skip to Content
Merck
All Photos(1)

Key Documents

MABE2044

Sigma-Aldrich

Anti-SPIB Antibody, clone 235D

Synonym(s):

Transcription factor Spi-B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

235D, monoclonal

mol wt

calculated mol wt 28.82 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

human ... SPIB(6689)

General description

Transcription factor Spi-B (UniProt: Q01892; also known as SPIB) is encoded by the SPIB gene (Gene ID: 6689) in human. SPIB is a member of the ETS family of transcription factors that is expressed in mature B cells, T-cell progenitors, and in plasmacytoid dendritic cells. It is reported to be a key regulator of human plasmacytoid dendritic cell development. It may also be required for B-cell receptor (BCR) signaling that is essential for normal B-cell development and antigenic stimulation. It serves as a sequence specific transcriptional activator that binds to the PU-box, a purine-rich DNA sequence (5′-GAGGAA-3′) that can act as a lymphoid-specific enhancer. SPIB contains a weakly acidic N-terminal transactivation domain (TAD; aa 1-31) that is followed by a second TAD (aa 41-61) that is rich in proline, serine, and threonine. Each of these domains may be required for transcriptional activation of a subset of target genes. Overexpression of SPIB is reported in all cases of blastic plasmacytoid dendritic cell (BPDC) neoplasms, but not in its cutaneous mimics. It can be used as a novel immunohistochemical marker for human BPDC neoplasm. (Ref.: Montes-Moreno, S., et al. (2013). Blood. 121(4); 643-647).

Specificity

Clone 235D is a mouse monoclonal antibody that detects Transcription factor Spi-B (SPIB).

Immunogen

His-tagged recombinant human transcription factor Spi-B (SPIB).

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human tonsil tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution of this antibody detected SPIB in human tonsil tissue sections.

Tested Applications

Western Blotting Analysis: A representative lot detected SPIB in Western Blotting application (Montes-Moreno, S., et al. (2013). Blood. 121(4); 643-7).

Immunohistochemistry (Paraffin) Analysis (IHC(P)): A representative lot detected SPIB in Immunohistochemistry application (Montes-Moreno, S., et al. (2013). Blood. 121(4); 643-7).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SPIB, clone 235D, Cat. No. MABE2044, is a mouse monoclonal antibody that detects transcription factor Spi-B (SPIB) and is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service