Skip to Content
Merck
All Photos(2)

Key Documents

ABD24

Sigma-Aldrich

Anti-Dermo-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Twist-related protein 2, Class A basic helix-loop-helix protein 39, bHLHa39, Dermis-expressed protein 1, Dermo-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

species reactivity (predicted by homology)

chicken (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TWIST2(117581)

General description

Dermo-1, also known as Twist-related protein 2, is a basic helix-loop-helix transcription factor that has been shown to be a negative regulator for gene transcription and apoptosis. Basic helix-loop-helix transcription factors are implicated in cell lineage determination and differentiation. Dermo-1 shares extensive homology with another basic helix-loop-helix transcription factor, Twist. Studies suggest that Dermo-1 may inhibit osteoblast maturation during osteoblast development and maintain cells in preosteoblast phenotype.

Immunogen

KLH-conjugated linear peptide corresponding to human Dermo-1.

Application

Anti-Dermo-1 Antibody is an antibody against Dermo-1 for use in WB, IH(P).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Dermo-1 in rat liver tissue.
Research Category
Stem Cell Research
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in rat liver tissue lysate.

Western Blot Analysis: 0.05 µg/mL of this antibody detected Dermo-1 on 10 µg of rat liver tissue lysate.

Target description

~18 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat liver tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service