Skip to Content
Merck
All Photos(1)

Key Documents

6.17797

Supelco

Fehling′s solution A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302

form

solution

Application


  • Chemical and enzymic analyses of Pichia polymorpha cell walls: This study demonstrates the use of Fehling′s solution A in biochemical research, specifically for the chemical and enzymatic analysis of yeast cell walls. The use of Fehling′s solution A as a copper complex reagent provides critical insights into the carbohydrate content of cell walls, which is essential for understanding cell wall structure and function in microbiology. This research highlights Fehling′s solution A′s application in detailed organic compound identification within a microbial context (Villa et al., 1980).

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service