Skip to Content
Merck
All Photos(1)

Key Documents

208736

Sigma-Aldrich

Calmodulin Inhibitory Peptide, Control

The Calmodulin Inhibitory Peptide, Control controls the biological activity of Calmodulin. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

Calmodulin Inhibitory Peptide, Control, MLCK Control Peptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C84H149N35O24
Molecular Weight:
2033.30
UNSPSC Code:
12352202

Quality Level

Assay

≥94% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A 17 amino acid peptide suitable for use as a control with Calmodulin Inhibitory Peptide (Cat. No. 208735).
A control peptide for Calmodulin Inhibitory Peptide (Cat. No. 208735).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A control peptide for Calmodulin Inhibitory Peptide (Cat. No. 208735)
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Arg-Arg-Lys-Glu-Gln-Lys-Thr-Gly-His-Ala-Val-Arg-Ala-Ile-Gly-Arg-Glu-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Torok, K. et al., 1998. Biochemistry.37, 6188.
Török, K., and Trentham, D.R. 1994. Biochemistry33, 12807.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service