Skip to Content
Merck
All Photos(1)

Key Documents

P75903

Sigma-Aldrich

3-Pyrroline

65%

Synonym(s):

2,5-Dihydropyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N
CAS Number:
Molecular Weight:
69.11
Beilstein:
103173
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

concentration

65%

refractive index

n20/D 1.46 (lit.)

bp

90-91 °C/748 mmHg (lit.)

density

0.91 g/mL at 25 °C (lit.)

SMILES string

C1NCC=C1

InChI

1S/C4H7N/c1-2-4-5-3-1/h1-2,5H,3-4H2

InChI key

JVQIKJMSUIMUDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Remainder pyrrolidine

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F

Flash Point(C)

-18 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service